The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053250.2 | NP_444480.1 | 4 - 5 | 337 | 108 |
NM_181664.2 | NP_858050.1 | 4 - 5 | 337 | 108 | |
GenBank mRNA | AF367970.1 | - | 4 - 5 | 337 | 108 |
AF367971.1 | - | 4 - 5 | 337 | 108 | |
AK080272.1 | - | 4 - 5 | 337 | 108 | |
AY555745.1 | - | 4 - 5 | 328 | 108 | |
BC116360.1 | - | 4 - 5 | 337 | 108 |