Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tle6
Entrez Gene ID
114606
Gene Name
transducin-like enhancer of split 6, homolog of Drosophila E(spl)
Gene Aliases
1810057E06Rik, Grg6
Chromosome Location
Chr.10: 81590904 - 81601190 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053254.2 NP_444484.1 7 - 8 868 70
GenBank mRNA AB221587.1 - 6 - 7 729 70
AF145957.2 - 7 - 8 868 70
AK135768.1 - 7 - 8 845 70
AK154391.1 - 4 - 5 828 70
BC048832.1 - 4 - 5 707 70

View Full Product Details