The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053260.3 | NP_444490.2 | 3 - 4 | 400 | 74 |
GenBank mRNA | AF305425.2 | - | 2 - 3 | 236 | 74 |
AF442819.1 | - | 2 - 3 | 236 | 74 | |
AK144368.1 | - | 3 - 4 | 401 | 74 | |
BC104122.1 | - | 2 - 3 | 236 | 74 | |
BC104123.1 | - | 2 - 3 | 236 | 74 |