Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Afg3l1
Entrez Gene ID
114896
Gene Name
AFG3-like AAA ATPase 1
Gene Aliases
1700047G05Rik, 3110061K15Rik
Chromosome Location
Chr.8: 123477862 - 123503916 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054070.3 NP_473411.2 16 - 17 2204 80
NR_028260.1 15 - 16 2088 80
GenBank mRNA AF329695.1 - 16 - 17 2145 80
AK006708.1 - 10 - 11 1475 80
AK012394.1 - 2 - 3 381 80
AK159647.1 - 16 - 17 2168 80
AK167964.1 - 16 - 17 2179 80
AK168244.1 - 15 - 16 2062 80
AK213495.1 - 1 - 2 77 80
BC043320.1 - 6 - 7 910 80
BC056978.1 - 16 - 17 2163 80
BC060064.1 - 15 - 16 2047 80

View Full Product Details