Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Optc
Entrez Gene ID
269120
Gene Name
opticin
Gene Aliases
-
Chromosome Location
Chr.1: 133897193 - 133909714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160422.1 NP_001153894.1 3 - 4 654 62
NM_054076.3 NP_473417.2 3 - 4 654 62
GenBank mRNA AF333980.1 - 3 - 4 653 62
AK053318.1 - 3 - 4 437 62
AY077682.1 - 3 - 4 646 62
BC099435.1 - 3 - 4 487 62

View Full Product Details