Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fubp1
Entrez Gene ID
51886
Gene Name
far upstream element (FUSE) binding protein 1
Gene Aliases
9530027K12Rik, D3Ertd330e, FBP, Fubp, Fubp4
Chromosome Location
Chr.3: 152210427 - 152236830 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057172.3 NP_476513.2 9 - 10 857 81
GenBank mRNA AK028418.1 - 9 - 10 858 81
AK029458.1 - 10 - 11 876 81
AK032517.1 - 9 - 10 824 81
AK138011.1 - 9 - 10 802 81
AK160815.1 - 9 - 10 795 81
AK166066.1 - 9 - 10 802 81
AK167730.1 - 9 - 10 837 81
BC014763.1 - 9 - 10 820 81
BC023780.1 - 9 - 10 838 81

View Full Product Details