The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057172.3 | NP_476513.2 | 16 - 17 | 1696 | 70 |
GenBank mRNA | AK028418.1 | - | 16 - 17 | 1698 | 70 |
AK138011.1 | - | 16 - 17 | 1641 | 70 | |
AK160815.1 | - | 16 - 17 | 1634 | 70 | |
AK166066.1 | - | 16 - 17 | 1641 | 70 | |
AK167730.1 | - | 16 - 17 | 1676 | 70 | |
BC014763.1 | - | 16 - 17 | 1659 | 70 | |
BC023780.1 | - | 16 - 17 | 1677 | 70 |