Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Strc
Entrez Gene ID
140476
Gene Name
stereocilin
Gene Aliases
DFNB16
Chromosome Location
Chr.2: 121363248 - 121381370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080459.2 NP_536707.2 5 - 6 2384 84
GenBank mRNA AF375593.1 - 5 - 6 2384 84

View Full Product Details