The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080558.4 | NP_542125.3 | 16 - 17 | 3762 | 76 |
GenBank mRNA | AB093303.1 | - | 16 - 17 | 3637 | 76 |
AB120565.1 | - | 16 - 17 | 3647 | 76 | |
AK037213.1 | - | 14 - 15 | 3472 | 76 | |
AK169543.1 | - | 17 - 18 | 3940 | 76 | |
BC008560.1 | - | 9 - 10 | 2735 | 76 | |
BC018526.1 | - | 6 - 7 | 966 | 76 | |
BC079535.1 | - | 10 - 11 | 3141 | 76 | |
BC115799.1 | - | 3 - 4 | 548 | 76 | |
BC122519.1 | - | 16 - 17 | 3648 | 76 | |
BC141883.1 | - | 16 - 17 | 3647 | 76 |