The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080575.2 | NP_542142.1 | 8 - 9 | 1361 | 59 |
GenBank mRNA | AB046742.1 | - | 8 - 9 | 1350 | 59 |
AK146568.1 | - | 8 - 9 | 1362 | 59 | |
AK158119.1 | - | 8 - 9 | 1356 | 59 | |
AK173275.1 | - | 8 - 9 | 1322 | 59 | |
BC030930.1 | - | 4 - 5 | 535 | 59 | |
BC092278.1 | - | 8 - 9 | 1390 | 59 |