Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acss1
Entrez Gene ID
68738
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
1110032O15Rik, AI788978, Acas2, Acas2l, AceCS2
Chromosome Location
Chr.2: 150618111 - 150668240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080575.2 NP_542142.1 9 - 10 1479 68
GenBank mRNA AB046742.1 - 9 - 10 1468 68
AK088244.1 - 9 - 10 1458 68
AK146568.1 - 9 - 10 1480 68
AK158119.1 - 9 - 10 1474 68
AK172412.1 - 9 - 10 1465 68
AK172470.1 - 9 - 10 1454 68
AK173275.1 - 9 - 10 1440 68
BC030930.1 - 5 - 6 653 68
BC092278.1 - 9 - 10 1508 68

View Full Product Details