The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102404.1 | NP_001095874.1 | 3 - 4 | 520 | 79 |
NM_001102405.1 | NP_001095875.1 | 3 - 4 | 525 | 79 | |
NM_007388.3 | NP_031414.1 | 3 - 4 | 467 | 79 | |
GenBank mRNA | AK010865.1 | - | 3 - 4 | 491 | 79 |
AK011915.1 | - | 3 - 4 | 491 | 79 | |
BC012911.1 | - | 3 - 4 | 419 | 79 | |
BC019160.1 | - | 3 - 4 | 483 | 79 | |
BC029644.1 | - | 3 - 4 | 419 | 79 | |
S70805.1 | - | 2 - 3 | 143 | 79 |