The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007405.2 | NP_031431.2 | 3 - 4 | 1158 | 82 |
GenBank mRNA | BU611374.1 | - | 2 - 3 | 362 | 82 |
M93422.1 | - | 2 - 3 | 1099 | 82 | |
M96653.1 | - | 2 - 3 | 986 | 82 |