Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap2a1
Entrez Gene ID
11771
Gene Name
adaptor-related protein complex 2, alpha 1 subunit
Gene Aliases
Adtaa
Chromosome Location
Chr.7: 44900373 - 44929490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077264.1 NP_001070732.1 12 - 13 1697 88
NM_007458.2 NP_031484.1 12 - 13 1697 88
GenBank mRNA AK135424.1 - 12 - 13 1626 88
AK155515.1 - 12 - 13 1634 88
AK198323.1 - 2 - 3 132 88
AK201691.1 - - 132 88
BC031433.1 - 12 - 13 1647 88

View Full Product Details