The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042527.2 | NP_001035992.1 | 18 - 19 | 3654 | 104 |
NM_007550.4 | NP_031576.4 | 18 - 19 | 3925 | 104 | |
GenBank mRNA | AB008674.1 | - | 18 - 19 | 3915 | 104 |
Z98263.1 | - | 18 - 19 | 3632 | 104 |