The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042527.2 | NP_001035992.1 | 19 - 20 | 3848 | 97 |
NM_007550.4 | NP_031576.4 | 19 - 20 | 4119 | 97 | |
GenBank mRNA | AB008674.1 | - | 19 - 20 | 4109 | 97 |
Z98263.1 | - | 19 - 20 | 3826 | 97 |