The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130443.1 | NP_001123915.1 | 4 - 5 | 657 | 74 |
NM_008284.2 | NP_032310.2 | 4 - 5 | 657 | 74 | |
GenBank mRNA | AY373386.1 | - | 3 - 4 | 451 | 74 |
BC011083.1 | - | 4 - 5 | 585 | 74 | |
BC061885.1 | - | - | 613 | 74 | |
BU924724.1 | - | 2 - 3 | 293 | 74 |