Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nbr1
Entrez Gene ID
17966
Gene Name
neighbor of Brca1 gene 1
Gene Aliases
mKIAA0049
Chromosome Location
Chr.11: 101552107 - 101581951 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252220.1 NP_001239149.1 14 - 15 2201 68
NM_001252222.1 NP_001239151.1 14 - 15 2024 68
NM_001252223.1 NP_001239152.1 14 - 15 2024 68
NM_008676.3 NP_032702.1 14 - 15 2024 68
GenBank mRNA AF227188.1 - 14 - 15 2153 68
AF408436.1 - 5 - 6 862 68
AK077479.1 - 4 - 5 809 68
AK128992.1 - 14 - 15 1816 68
AK129043.1 - 14 - 15 1794 68
AK154081.1 - 14 - 15 1810 68
BC052709.1 - 14 - 15 1814 68
BC129867.1 - 13 - 14 1676 68
BC129868.1 - 13 - 14 1676 68
U73039.1 - 14 - 15 1820 68

View Full Product Details