The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008682.2 | NP_032708.2 | 8 - 9 | 1224 | 122 |
GenBank mRNA | AK010591.1 | - | 8 - 9 | 1212 | 122 |
AK048393.1 | - | 8 - 9 | 122 | ||
AK085355.1 | - | 8 - 9 | 1187 | 122 | |
AK090040.1 | - | 8 - 9 | 1169 | 122 | |
BC052430.1 | - | 8 - 9 | 1193 | 122 | |
BC066870.1 | - | 8 - 9 | 1161 | 122 | |
D10712.1 | - | 8 - 9 | 1166 | 122 |