The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008683.3 | NP_032709.1 | 1 - 2 | 117 | 77 |
GenBank mRNA | AK146732.1 | - | 1 - 2 | 118 | 77 |
AK147080.1 | - | 1 - 2 | 102 | 77 | |
BC004625.1 | - | 1 - 2 | 102 | 77 | |
D10918.1 | - | 1 - 2 | 84 | 77 |