Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nxn
Entrez Gene ID
18230
Gene Name
nucleoredoxin
Gene Aliases
l11Jus13
Chromosome Location
Chr.11: 76257226 - 76399141 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008750.5 NP_032776.1 5 - 6 904 59
GenBank mRNA AB095441.1 - 5 - 6 749 59
AB095442.1 - 5 - 6 749 59
AB095443.1 - 5 - 6 749 59
AB095444.1 - 5 - 6 749 59
AK012352.1 - 5 - 6 889 59
AK048109.1 - 5 - 6 904 59
AK149075.1 - 5 - 6 888 59
AK192350.1 - 1 - 2 59
BC004688.1 - 5 - 6 864 59
BC047339.1 - 4 - 5 436 59
BC058244.1 - 5 - 6 904 59

View Full Product Details