The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281934.1 | NP_001268863.1 | 16 - 17 | 2860 | 72 |
NM_008794.2 | NP_032820.2 | 16 - 17 | 2596 | 72 | |
NR_104055.1 | 17 - 18 | 2671 | 72 | ||
GenBank mRNA | AK148416.1 | - | 16 - 17 | 2181 | 72 |
AK151980.1 | - | 16 - 17 | 2183 | 72 | |
AK165389.1 | - | 16 - 17 | 2202 | 72 | |
AK207794.1 | - | 1 - 2 | 80 | 72 | |
BC006730.1 | - | 15 - 16 | 2097 | 72 | |
BC007322.1 | - | 16 - 17 | 2457 | 72 | |
BC056456.1 | - | 17 - 18 | 2670 | 72 | |
U48830.1 | - | 16 - 17 | 2168 | 72 |