The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008818.2 | NP_032844.2 | 3 - 4 | 603 | 59 |
GenBank mRNA | AK146379.1 | - | 3 - 4 | 628 | 59 |
AK146393.1 | - | 3 - 4 | 627 | 59 | |
AK146449.1 | - | 3 - 4 | 609 | 59 | |
BC056204.1 | - | 4 - 5 | 611 | 59 | |
DQ058642.1 | - | 3 - 4 | 587 | 59 | |
M32484.1 | - | 3 - 4 | 587 | 59 |