The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008849.4 | NP_032875.1 | 2 - 3 | 218 | 75 |
GenBank mRNA | AB622207.1 | - | 1 - 2 | 362 | 75 |
BC061213.1 | - | - | 335 | 75 | |
D12885.1 | - | - | 336 | 75 | |
X57512.1 | - | 2 - 3 | 218 | 75 |