The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008916.2 | NP_032942.1 | 1 - 2 | 179 | 58 |
GenBank mRNA | AF483522.1 | - | 1 - 2 | 99 | 58 |
AF483523.1 | - | 1 - 2 | 99 | 58 | |
AK052817.1 | - | 1 - 2 | 171 | 58 | |
AK054436.1 | - | 1 - 2 | 198 | 58 | |
AK164290.1 | - | 1 - 2 | 168 | 58 | |
BC066112.1 | - | 1 - 2 | 179 | 58 | |
U96724.1 | - | 1 - 2 | 210 | 58 |