Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptx3
Entrez Gene ID
19288
Gene Name
pentraxin related gene
Gene Aliases
AI607804, TSG-14
Chromosome Location
Chr.3: 66219887 - 66225806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008987.3 NP_033013.3 1 - 2 306 141
GenBank mRNA BC022176.1 - 1 - 2 306 141
BQ442682.1 - 1 - 2 193 141
CA529293.1 - 1 - 2 306 141
X83601.1 - 1 - 2 230 141

View Full Product Details