The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009682.3 | NP_033812.3 | 2 - 3 | 226 | 85 |
GenBank mRNA | AK028372.1 | - | 2 - 3 | 209 | 85 |
AK037336.1 | - | 2 - 3 | 204 | 85 | |
AK083911.1 | - | 2 - 3 | 215 | 85 | |
AK150085.1 | - | 2 - 3 | 219 | 85 | |
BC060236.1 | - | 2 - 3 | 187 | 85 | |
BY127127.1 | - | 2 - 3 | 229 | 85 | |
U91933.1 | - | 2 - 3 | 181 | 85 |