The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009705.3 | NP_033835.1 | 3 - 4 | 447 | 69 |
GenBank mRNA | AF032466.1 | - | 3 - 4 | 399 | 69 |
BC023349.1 | - | 3 - 4 | 411 | 69 | |
DV039756.1 | - | 3 - 4 | 481 | 69 | |
U90886.1 | - | 3 - 4 | 419 | 69 |