Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nf2
Entrez Gene ID
18016
Gene Name
neurofibromatosis 2
Gene Aliases
merlin
Chromosome Location
Chr.11: 4757650 - 4849544 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252250.1 NP_001239179.1 2 - 3 802 65
NM_001252251.1 NP_001239180.1 2 - 3 802 65
NM_001252252.1 NP_001239181.1 2 - 3 802 65
NM_001252253.1 NP_001239182.1 2 - 3 267 65
NM_010898.4 NP_035028.2 2 - 3 802 65
GenBank mRNA AK017343.1 - 2 - 3 268 65
AK045998.1 - 2 - 3 624 65
AK185632.1 - 1 - 2 90 65
AK213371.1 - 1 - 2 90 65
BC005442.1 - 2 - 3 668 65
CN715904.1 - 2 - 3 360 65
L22987.1 - 2 - 3 260 65
L22988.1 - 2 - 3 260 65
L22989.1 - 2 - 3 260 65
L27090.1 - 2 - 3 253 65
L27105.1 - 2 - 3 253 65
L28176.1 - 2 - 3 560 65
X74671.1 - 2 - 3 819 65

View Full Product Details