The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037177.2 | NP_001032254.1 | 2 - 3 | 1336 | 52 |
NM_001037178.2 | NP_001032255.1 | 2 - 3 | 701 | 52 | |
NM_001136073.2 | NP_001129545.1 | 2 - 3 | 1336 | 52 | |
NM_001291168.1 | NP_001278097.1 | 2 - 3 | 1221 | 52 | |
NM_001291169.1 | NP_001278098.1 | 2 - 3 | 1221 | 52 | |
NM_001291170.1 | NP_001278099.1 | 2 - 3 | 701 | 52 | |
NM_001291172.1 | NP_001278101.1 | 2 - 3 | 1221 | 52 | |
NM_001291174.1 | NP_001278103.1 | 2 - 3 | 701 | 52 | |
NM_001291175.1 | NP_001278104.1 | 2 - 3 | 816 | 52 | |
NM_001291177.1 | NP_001278106.1 | 2 - 3 | 816 | 52 | |
NM_001291179.1 | NP_001278108.1 | 2 - 3 | 816 | 52 | |
NM_010899.3 | NP_035029.2 | 2 - 3 | 1336 | 52 | |
GenBank mRNA | AF289078.1 | - | 2 - 3 | 1162 | 52 |
AK044489.1 | - | 2 - 3 | 1222 | 52 | |
AK081853.1 | - | 2 - 3 | 1304 | 52 | |
AK083119.1 | - | 2 - 3 | 677 | 52 | |
AK135414.1 | - | 2 - 3 | 1338 | 52 | |
BC064006.1 | - | 2 - 3 | 1255 | 52 | |
EU887581.1 | - | 2 - 3 | 1102 | 52 | |
EU887582.1 | - | 2 - 3 | 1162 | 52 | |
EU887583.1 | - | 2 - 3 | 699 | 52 | |
EU887584.1 | - | 2 - 3 | 799 | 52 | |
EU887587.1 | - | 2 - 3 | 1102 | 52 | |
EU887588.1 | - | 2 - 3 | 1162 | 52 | |
EU887589.1 | - | 2 - 3 | 699 | 52 | |
EU887590.1 | - | 2 - 3 | 799 | 52 | |
EU887593.1 | - | 2 - 3 | 1102 | 52 | |
EU887594.1 | - | 2 - 3 | 1162 | 52 | |
EU887595.1 | - | 2 - 3 | 699 | 52 | |
EU887596.1 | - | 2 - 3 | 799 | 52 | |
EU887599.1 | - | 2 - 3 | 1102 | 52 | |
EU887600.1 | - | 2 - 3 | 1162 | 52 | |
EU887601.1 | - | 2 - 3 | 699 | 52 | |
EU887602.1 | - | 2 - 3 | 799 | 52 | |
U36575.1 | - | 2 - 3 | 1232 | 52 | |
U36576.1 | - | 2 - 3 | 1232 | 52 |