Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Numbl
Entrez Gene ID
18223
Gene Name
numb-like
Gene Aliases
nbl
Chromosome Location
Chr.7: 27258438 - 27282150 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010950.2 NP_035080.2 9 - 10 1339 129
GenBank mRNA AK147523.1 - 7 - 8 7126 129
BC059915.1 - 8 - 9 1928 129
BC068116.1 - 9 - 10 1166 129
BC098097.1 - 9 - 10 1180 129
U96441.1 - 10 - 11 1220 129

View Full Product Details