Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pygm
Entrez Gene ID
19309
Gene Name
muscle glycogen phosphorylase
Gene Aliases
AI115133, PG
Chromosome Location
Chr.19: 6384429 - 6398459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011224.1 NP_035354.1 5 - 6 708 61
GenBank mRNA AF124787.1 - 5 - 6 708 61
BC012961.1 - 5 - 6 714 61

View Full Product Details