Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pygm
Entrez Gene ID
19309
Gene Name
muscle glycogen phosphorylase
Gene Aliases
AI115133, PG
Chromosome Location
Chr.19: 6384429 - 6398459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011224.1 NP_035354.1 18 - 19 2359 104
GenBank mRNA AF124787.1 - 18 - 19 2359 104
AK019077.1 - 1 - 2 138 104
BC012961.1 - 18 - 19 2365 104

View Full Product Details