Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apc2
Entrez Gene ID
23805
Gene Name
adenomatosis polyposis coli 2
Gene Aliases
AI852447, APCL, R75424
Chromosome Location
Chr.10: 80295957 - 80318257 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011789.2 NP_035919.2 13 - 14 1845 74

View Full Product Details