The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016709.2 | NP_057918.2 | 4 - 5 | 462 | 69 |
GenBank mRNA | AK003219.1 | - | 4 - 5 | 441 | 69 |
AK134886.1 | - | 4 - 5 | 434 | 69 | |
BC026525.1 | - | 4 - 5 | 400 | 69 | |
BC049597.1 | - | 4 - 5 | 446 | 69 |