Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bok
Entrez Gene ID
51800
Gene Name
BCL2-related ovarian killer
Gene Aliases
AI847676, AU021146, matador, mtd
Chromosome Location
Chr.1: 93685694 - 93695762 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016778.2 NP_058058.1 2 - 3 408 67
GenBank mRNA AF027707.1 - 2 - 3 406 67
AK156243.1 - 2 - 3 443 67
AK168373.1 - 2 - 3 339 67
AK171258.1 - 2 - 3 365 67
BC030069.1 - 2 - 3 346 67

View Full Product Details