The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016778.2 | NP_058058.1 | 3 - 4 | 544 | 84 |
GenBank mRNA | AF027707.1 | - | 3 - 4 | 542 | 84 |
AK156243.1 | - | 3 - 4 | 579 | 84 | |
AK168373.1 | - | 3 - 4 | 475 | 84 | |
AK171258.1 | - | 3 - 4 | 501 | 84 | |
BC030069.1 | - | 3 - 4 | 482 | 84 |