Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nphp1
Entrez Gene ID
53885
Gene Name
nephronophthisis 1 (juvenile) homolog (human)
Gene Aliases
-
Chromosome Location
Chr.2: 127740732 - 127788914 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291012.1 NP_001277941.1 1 - 2 103 79
NM_001291013.1 NP_001277942.1 1 - 2 103 79
NM_016902.4 NP_058598.1 1 - 2 103 79
GenBank mRNA AF127180.1 - 1 - 2 66 79
AF176210.1 - 1 - 2 66 79
AJ243223.1 - 1 - 2 66 79
AK159627.1 - 1 - 2 80 79
BC118953.2 - 1 - 2 66 79
BY147655.1 - 1 - 2 104 79
EF174553.3 - 1 - 2 75 79
EF174554.2 - 1 - 2 75 79
FJ767895.1 - 1 - 2 66 79
FJ767896.1 - 1 - 2 66 79
FJ767897.1 - 1 - 2 66 79
FJ767898.1 - 1 - 2 66 79

View Full Product Details