Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nphp1
Entrez Gene ID
53885
Gene Name
nephronophthisis 1 (juvenile) homolog (human)
Gene Aliases
-
Chromosome Location
Chr.2: 127740732 - 127788914 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291012.1 NP_001277941.1 16 - 17 1604 72
NM_001291013.1 NP_001277942.1 16 - 17 1601 72
NM_016902.4 NP_058598.1 16 - 17 1607 72
GenBank mRNA AF127180.1 - 16 - 17 1558 72
AF176210.1 - 16 - 17 1570 72
AJ243223.1 - 16 - 17 1555 72
AK159627.1 - 16 - 17 1581 72
BC118953.2 - 16 - 17 1552 72
EF174553.3 - 16 - 17 1576 72

View Full Product Details