The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017374.3 | NP_059070.1 | 2 - 3 | 611 | 94 |
GenBank mRNA | AK090311.1 | - | 2 - 3 | 244 | 94 |
BC019161.1 | - | 2 - 3 | 94 | ||
BC049594.1 | - | 2 - 3 | 380 | 94 | |
BC058582.1 | - | 2 - 3 | 94 | ||
Z67746.1 | - | 2 - 3 | 315 | 94 |