Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 6 - 7 2661 95
GenBank mRNA AF096286.1 - 1 - 2 691 95
AK129221.1 - 1 - 2 541 95
BC066849.1 - 1 - 2 349 95
BC098370.1 - 1 - 2 266 95

View Full Product Details