Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1e1
Entrez Gene ID
56043
Gene Name
aldo-keto reductase family 1, member E1
Gene Aliases
1810061I10Rik, Akr1e2
Chromosome Location
Chr.13: 4592490 - 4609164 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018859.2 NP_061347.2 4 - 5 450 66
GenBank mRNA AK002507.1 - 4 - 5 425 66
AK007924.1 - 4 - 5 427 66
AK028605.1 - - 276 66
BC012692.1 - 4 - 5 466 66
U68535.1 - 4 - 5 436 66

View Full Product Details