Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exosc9
Entrez Gene ID
50911
Gene Name
exosome component 9
Gene Aliases
PM/Scl-75, Pmscl1, RRP45, p5, p6
Chromosome Location
Chr.3: 36552606 - 36565727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019393.2 NP_062266.1 2 - 3 240 80
GenBank mRNA AF152841.1 - 2 - 3 213 80
AF152842.1 - 2 - 3 262 80
AK010253.1 - 2 - 3 215 80
AK011636.1 - 2 - 3 191 80
AK076096.1 - 2 - 3 222 80
AK083794.1 - 1 - 2 479 80
BC005622.1 - 2 - 3 219 80
BC052156.1 - 2 - 3 212 80
BC158075.1 - 2 - 3 191 80
BY078098.1 - 2 - 3 241 80

View Full Product Details