The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164187.1 | NP_001157659.1 | 9 - 10 | 1031 | 105 |
NM_019542.2 | NP_062415.1 | 9 - 10 | 1040 | 105 | |
GenBank mRNA | AJ242909.1 | - | 9 - 10 | 1052 | 105 |
AJ242911.1 | - | 9 - 10 | 992 | 105 | |
AK007240.1 | - | 9 - 10 | 1035 | 105 | |
AK155292.1 | - | 9 - 10 | 932 | 105 | |
BC004689.1 | - | 9 - 10 | 918 | 105 |