The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284328.1 | NP_001271257.1 | 9 - 10 | 1509 | 112 |
NM_019666.2 | NP_062640.2 | 9 - 10 | 1509 | 112 | |
NM_019796.5 | NP_062770.1 | 9 - 10 | 1509 | 112 | |
GenBank mRNA | AB035725.1 | - | 9 - 10 | 1308 | 112 |
AF093821.1 | - | 8 - 9 | 1170 | 112 | |
AF408434.1 | - | 7 - 8 | 962 | 112 | |
AK034845.1 | - | 9 - 10 | 1321 | 112 | |
AK076026.1 | - | 9 - 10 | 1299 | 112 | |
AK077588.1 | - | 9 - 10 | 1258 | 112 | |
AK078158.1 | - | 7 - 8 | 942 | 112 | |
AK083398.1 | - | 8 - 9 | 1100 | 112 | |
AK162448.1 | - | 3 - 4 | 391 | 112 | |
BC041148.1 | - | 9 - 10 | 1354 | 112 | |
BC050079.1 | - | 9 - 10 | 1282 | 112 | |
BC055863.1 | - | 9 - 10 | 1287 | 112 | |
BC058807.1 | - | 9 - 10 | 1376 | 112 | |
BC080309.1 | - | 9 - 10 | 1212 | 112 | |
BC108363.1 | - | 9 - 10 | 1243 | 112 |