The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019798.5 | NP_062772.3 | 6 - 7 | 1136 | 65 |
NM_183408.3 | NP_899668.1 | 11 - 12 | 1644 | 65 | |
GenBank mRNA | AB154250.1 | - | 6 - 7 | 728 | 65 |
AF208021.2 | - | 6 - 7 | 808 | 65 | |
AJ297396.1 | - | 6 - 7 | 808 | 65 | |
AK153657.1 | - | 11 - 12 | 1649 | 65 | |
AK160843.1 | - | 10 - 11 | 1078 | 65 | |
BC027224.1 | - | 11 - 12 | 1340 | 65 | |
BC044786.2 | - | 10 - 11 | 2206 | 65 | |
BC139405.1 | - | 11 - 12 | 1484 | 65 | |
BC139408.1 | - | 11 - 12 | 1484 | 65 |