The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020487.4 | NP_065233.2 | 3 - 4 | 317 | 80 |
GenBank mRNA | AB033822.1 | - | 3 - 4 | 303 | 80 |
AB059415.1 | - | 3 - 4 | 315 | 80 | |
AF176209.2 | - | 3 - 4 | 317 | 80 | |
AK006271.1 | - | 3 - 4 | 289 | 80 | |
AY005145.1 | - | 3 - 4 | 294 | 80 | |
BC049588.1 | - | 3 - 4 | 331 | 80 | |
BC116755.1 | - | 3 - 4 | 315 | 80 | |
BC116757.1 | - | 3 - 4 | 315 | 80 |