The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020579.2 | NP_065604.2 | 6 - 7 | 1103 | 95 |
GenBank mRNA | AF142671.1 | - | 6 - 7 | 1043 | 95 |
AK155471.1 | - | 7 - 8 | 1203 | 95 | |
BC004095.1 | - | 6 - 7 | 860 | 95 | |
BC013619.1 | - | 6 - 7 | 1076 | 95 | |
BC021830.1 | - | 4 - 5 | 1899 | 95 |