The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020581.2 | NP_065606.2 | 1 - 2 | 509 | 63 |
GenBank mRNA | AB054540.1 | - | 1 - 2 | 395 | 63 |
AF123261.1 | - | 1 - 2 | 440 | 63 | |
AF169313.1 | - | 1 - 2 | 372 | 63 | |
AF278699.1 | - | 1 - 2 | 444 | 63 | |
AK009161.1 | - | 1 - 2 | 215 | 63 | |
AK014564.1 | - | - | 510 | 63 | |
AK132761.1 | - | 1 - 2 | 478 | 63 | |
AK134419.1 | - | 1 - 2 | 478 | 63 | |
BC006611.1 | - | 1 - 2 | 461 | 63 | |
BC021343.1 | - | 1 - 2 | 475 | 63 | |
BC025797.1 | - | 1 - 2 | 445 | 63 | |
KJ650202.1 | - | 1 - 2 | 326 | 63 |