Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vstm2b
Entrez Gene ID
58188
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
2900093B09Rik, AB030198
Chromosome Location
Chr.7: 40898297 - 40930044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021387.3 NP_067362.1 4 - 5 861 70
GenBank mRNA AB030198.1 - 4 - 5 862 70
AK013845.1 - 4 - 5 848 70

View Full Product Details